Learn More
LOXL2 Mouse anti-Human, Unconjugated, Clone: 1H7F1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67139-1-IG-150UL
 View more versions of this product
Product Code. 16879595
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
The LOXL2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human LOXL2. This antibody recognizes human antigen. The LOXL2 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Specifications
LOXL2
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunofluorescence, ELISA
1H7F1
Unconjugated
150 μL
4017
IgG2a
Certificates
Promotions
Promotions
Product Content Correction
Product Title
LOXL2 Mouse anti-Human, Unconjugated, Clone: 1H7F1, Proteintech > 150 μL