Learn More
LOXL2 Mouse anti-Human, Unconjugated, Clone: 1H7F1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67139-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The LOXL2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human LOXL2. This antibody recognizes human antigen. The LOXL2 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
LOXL2 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
ELISA, Immunofluorescence, Western Blot | |
1H7F1 | |
Unconjugated | |
20 μL | |
4017 | |
IgG2a |