missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC29 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25664-1-AP-20UL
This item is not returnable.
View return policy
Specifications
LRRC29 | |
Polyclonal | |
Unconjugated | |
LRRC29 | |
F box protein FBL9, F box/LRR repeat protein 9, FBL9, FBXL9, LRRC29 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
234684, 26231 | |
-20°C | |
Liquid |
Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8WV35 | |
Lrrc29 | |
LRRC29 Fusion Protein Ag22577 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
LRRC29 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction