missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC29 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1706.43 SEK - 4863.32 SEK
Specifications
Antigen | LRRC29 |
---|---|
Concentration | 0.37 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Specifications
LRRC29 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
Q8WV35 | |
234684, 26231 | |
LRRC29 Fusion Protein Ag22577 | |
Primary | |
-20°C | |
LRRC29 |
0.37 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
F box protein FBL9, F box/LRR repeat protein 9, FBL9, FBXL9, LRRC29 | |
Lrrc29 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
LRRC29 Rabbit anti-Human, Mouse, Polyclonal, Proteintech