missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRTM3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27003-1-AP-150UL
This item is not returnable.
View return policy
Description
The Leucine-rich repeat transmembrane neuronal proteins (LRRTMs) are differentially expressed in the nervous system and were recently found to instruct presynaptic and mediate postsynaptic glutamatergic differentiation, with LRRTM1 and LRRTM2 most potent at inducing presynaptic differentiation. Recent evidence suggests that LRRTM3 promotes the processing of amyloid-precursor protein (APP) by the beta-secretase BACE, leading to the proteolytic production of the A-beta42 peptide that is the main component of amyloid plaques. Furthermore, LRRTM3 maps to a region of chromosome 10 linked to both late-onset Alzheimer′s disease (AD) and elevated plasma A-beta42 levels, suggesting that LRRTM3 is a functional and positional candidate gene for AD.Specifications
LRRTM3 | |
Polyclonal | |
Unconjugated | |
LRRTM3 | |
LRRTM3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
216028, 294380, 347731 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
D3ZAL8, Q86VH5, Q8BZ81 | |
LRRTM3 | |
LRRTM3 Fusion Protein Ag25452 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
LRRTM3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction