Learn More
luciferase Mouse anti-Gaussia Princeps, Unconjugated, Clone: 2D2E1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67293-1-IG-150UL
 View more versions of this product
Product Code. 16879835
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The luciferase antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of luciferase. This antibody recognizes gaussia princeps antigen. The luciferase antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
luciferase
Monoclonal
1000 ÎĽg/mL
Mouse
Primary
IgG2a
Western Blot
2D2E1
Unconjugated
150 ÎĽL
Plant
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
luciferase Mouse anti-Gaussia Princeps, Unconjugated, Clone: 2D2E1, Proteintech > 150 ÎĽL