Learn More
luciferase Mouse anti-Gaussia Princeps, Unconjugated, Clone: 2D2E1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67293-1-IG-20UL
 View more versions of this product
Product Code. 16889835
  • 1645.54 SEK / 20µL
Description
The luciferase antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of luciferase. This antibody recognizes gaussia princeps antigen. The luciferase antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
luciferase
Monoclonal
1000 ÎĽg/mL
Mouse
Primary
IgG2a
Western Blot
2D2E1
Unconjugated
20 ÎĽL
Plant
Certificates
Promotions
Promotions
Product Content Correction
Product Title
luciferase Mouse anti-Gaussia Princeps, Unconjugated, Clone: 2D2E1, Proteintech > 20 ÎĽL