missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP4K5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12827-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MAP4K5 may play a role in the response to environmental stress. Appears to act upstream of the JUN N-terminal pathway. This gene encodes a member of the serine/threonine protein kinase family, that is highly similar to yeast SPS1/STE20 kinase. Yeast SPS1/STE20 functions near the beginning of the MAP kinase signal cascades that is essential for yeast pheromone response. This kinase was shown to activate Jun kinase in mammalian cells, which suggested a role in stress response. Two alternatively spliced transcript variants encoding the same protein have been described for this gene.Especificaciones
MAP4K5 | |
Polyclonal | |
Unconjugated | |
MAP4K5 | |
GCKR, KHS, KHS1, MAP4K5, MAPKKKK5, MEK kinase kinase 5, MEKKK 5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11183, 399510, 503027 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BPM2, Q9Y4K4 | |
Map4k5 | |
MAP4K5 Fusion Protein Ag3547 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Sugerencias de productos
Customers who viewed this item also viewed.
Viewing 1-4 of
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
MAP4K5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido