missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBD5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15961-1-AP-150UL
This item is not returnable.
View return policy
Description
MBD5 binds to heterochromatin but does not interact with either methylated or unmethylated DNA (in vitro).Specifications
MBD5 | |
Polyclonal | |
Unconjugated | |
MBD5 | |
KIAA1461, MBD5, MRD1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
109241, 311026, 55777 | |
-20°C | |
Liquid |
Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
B1AYB6, Q9P267 | |
MBD5 | |
MBD5 Fusion Protein Ag8728 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MBD5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction