missing translation for 'onlineSavingsMsg'
Learn More

MBIP Mouse anti-Human, Clone: 2E6H11, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66102-1-IG-20UL

 View more versions of this product

Product Code. 16827675

  • 1645.54 SEK / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MBIP inhibits the MAP3K12 activity to induce the activation of the JNK/SAPK pathway. Component of the ATAC complex, a complex with histone acetyltransferase activity on histones H3 and H4.
Specifications

Specifications

MBIP
Monoclonal
1.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9NS73
MBIP
MBIP Fusion Protein Ag18090
20 μL
Primary
Human
Antibody
IgG2b
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
2E6H11
Unconjugated
MBIP
MBIP, MUK binding inhibitory protein
Mouse
Protein A
RUO
51562
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MBIP Mouse anti-Human, Clone: 2E6H11, Proteintech > 20 μL; Unconjugated

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.