missing translation for 'onlineSavingsMsg'
Learn More

MBL™ Class A Kohlrausch Flask

Expanded neck for easy introduction of viscous liquids or solid samples.

Brand:  MBL FRE011

 View more versions of this product

Product Code. 15567900

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Expanded neck for easy introduction of viscous liquids or solid samples.

  • Complies with BS 615
  • With expanded neck for easy introduction of viscous liquids or solid samples
  • 2 sizes available: 100mL for sugar analysis, 200mL for determination of oil viscosity by Engler method
  • Calibrated to Class A tolerances
  • White enamel inscriptions
  • Manufactured from soda-lime glass
TRUSTED_SUSTAINABILITY
Specifications

Specifications

100 mL
155 mm
Kohlrausch flask
Borosilicate Glass
2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MBL™ Class A Kohlrausch Flask > Capacity: 100mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.