Learn More
MED17 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11505-1-AP-150UL
 View more versions of this product
Product Code. 16828633
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The activation of gene transcription is a multistep process that is triggered by factors that recognize transcriptional enhancer sites in DNA. These factors work with co-activators to direct transcriptional initiation by the RNA polymerase II apparatus. The protein encoded by CRSP6 is a subunit of the CRSP (cofactor required for SP1 activation) complex, which, along with TFIID, is required for efficient activation by SP1. This protein is also a component of other multisubunit complexes e. g. thyroid hormone receptor-(TR-) associated proteins which interact with TR and facilitate TR function on DNA templates in conjunction with initiation factors and cofactors.
Specifications
Specifications
MED17
Polyclonal
Unconjugated
MED17
ARC77, CRSP complex subunit 6, CRSP6, CRSP77, DRIP77, DRIP80, MED17, mediator complex subunit 17, TRAP80
Rabbit
Antigen Affinity Chromatography
RUO
234959, 300367, 9440
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8VCD5, Q9NVC6
MED17
MED17 Fusion Protein Ag2057
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MED17 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated