Learn More
MED4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15951-1-AP-150UL
 View more versions of this product
Product Code. 16885354
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The Mediator is a multiprotein coactivator that is required by DNA-binding transcription factors for activation of polymerase II (see MIM 180660)-transcribed genes. MED4 appears to be a core Mediator subunit and is found in nearly all Mediator preparations (summary by Sato et al., 2004).
Specifications
Specifications
MED4
Polyclonal
Unconjugated
MED4
ARC36, DRIP36, HSPC126, MED4, mediator complex subunit 4, RP11 90M2.2, TRAP36, VDRIP
Rabbit
Antigen Affinity Chromatography
RUO
29079, 306030, 67381
-20°C
Liquid
Western Blot
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q561Q8, Q9CQA5, Q9NPJ6
MED4
MED4 Fusion Protein Ag8629
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MED4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated