Learn More
Sampling Systems™ Metal Detectable Long Handled Spoons
Metal Detectable Long Handled Spoons with long handles make taking powder and liquid samples easy. They can also be used for general production work. The special metal detectable PP means the material will be picked up by a ray and general detection equipment.
Brand:  Sampling Systems™ M1036B-20
Product Code. 17594716
  • 179.00 SEK / Pack of 4
Only null left
Estimated Shipment: 24-07-2025
Description
  • Powder sampling is easy with our metal detectable spoons.
  • They have a very long handle which means they are easy to see if left in the powder.
  • They are made from a special robust, chemically resistant grade of PP.
  • The whole range is manufactured in cleanroom conditions and packed in a class 7 (10,000) cleanroom.
  • This ensures the cleanliness of the product.
  • The robust design means they can be used for years and years or used as a single use product straight from the bag.
  • Simply unwrap and use.
  • They are ideal for use with liquids, creams and pastes.
  • The blue color means they can easily seen if left in the material in line with HACCP guidelines.
  • Made from high quality PP.
  • Conforms to the latest FDA and EU food requirements.
  • 20 mL volume is a standard size.
Specifications
Specifications
Polypropylene
20 mL
Blue
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number065818 Certificate TypeCertificate of Compliance Date01/03/2023 Catalog NumberM1036B-20
Lot Number17594716 Certificate TypeCertificate of Compliance Date22/02/2023 Catalog NumberM1036B-20
Promotions
Promotions
Product Content Correction
Product Title
Sampling Systems™ Metal Detectable Long Handled Spoons > Metal Detectable Long Handled Spoons; Capacity (Metric): 20 mL; Color: Blue; Material: Polypropylene