missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanesulfonic anhydride, 97%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 69.0°C to 70.0°C |
Color | Gray to White |
Boiling Point | 125.0°C (4.0 mmHg) |
Infrared Spectrum | Authentic |
Assay Percent Range | 97% |
Packaging | Glass bottle |
Quantity | 5 g |
Synonym | methanesulfonic anhydride, methanesulfonic acid, anhydride, mesyl anhydride, mesic anhydride, methanesulphonic anhydride, methane sulfonic acid anhydride, methanesulfonyl methanesulfonate, methane sulfonic anhydride, methanesulfonic acid, 1,1'-anhydride, methanesulfonic acid methylsulfonyl ester |
Solubility Information | Solubility in water: reacts |
SMILES | CS(=O)(=O)OS(=O)(=O)C |
Show More |
Safety and Handling

Product Identifier
- Methanesulfonic anhydride
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- EUH014-Reacts violently with water.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Methanesulfonic anhydride, 97% >
Spot an opportunity for improvement?Share a Content Correction