Learn More
SP Bel-Art™ Metric Color Octagon Spinbar Magnetic Stirring Bars
Interrupted profile provides greater surface area and added turbulence when compared to the smooth surface of cylindrical bars
Brand: SP Bel-Art™ F37107-5108
This item is not returnable.
View return policy
Description
- The most commonly used shape.
- Choose from red, yellow or blue.
- Pivot ring allows the Spin Bar to “rock” while spinning for better mixing conditions
Specifications
Autoclavable | |
PTFE, FDA-grade, USP Class VI | |
51 mm | |
5 Pk. |
Red | |
8 mm | |
PTFE |
Product Content Correction
Product Title
SP Bel-Art™ Metric Color Octagon Spinbar Magnetic Stirring Bars > 51 x 8 mm, Red
Spot an opportunity for improvement?Share a Content Correction