Learn More
SP Bel-Art™ Metric Color Octagon Spinbar Magnetic Stirring Bars
Interrupted profile provides greater surface area and added turbulence when compared to the smooth surface of cylindrical bars
Brand:  SP Bel-Art™ F37107-5108
 View more versions of this product
Product Code. 16885373
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
  • The most commonly used shape.
  • Choose from red, yellow or blue.
  • Pivot ring allows the Spin Bar to “rock” while spinning for better mixing conditions
Specifications
Specifications
Autoclavable
PTFE, FDA-grade, USP Class VI
51 mm
5 Pk.
Red
8 mm
PTFE
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SP Bel-Art™ Metric Color Octagon Spinbar Magnetic Stirring Bars > 51 x 8 mm, Red