missing translation for 'onlineSavingsMsg'
Learn More

SP Bel-Art™ Metric Micro Spinbar Magnetic Stirring Bars

Designed for small volume mixing in vessels such as vials, tubes and gradient makers.

Brand:  SP Bel-Art™ F37119-0603

 View more versions of this product

Product Code. 16885403

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

  • Particularly useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE Coating - Superior Chemical Compatibility
  • Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
  • Autoclave safe
Specifications

Specifications

Autoclavable
PTFE, FDA-grade, USP Class VI
6 mm
10 Pk.
White
3 mm
PTFE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
SP Bel-Art™ Metric Micro Spinbar Magnetic Stirring Bars > 6 x 3 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.