Learn More
SP Bel-Art™ Metric Polygon Spinbar Magnetic Stirring Bars without Pivot Ring
High quality PTFE encapsulated magnetic stirring bars ensure efficient mixing with consistent results.
Brand:  SP Bel-Art™ F37120-3810
 View more versions of this product
Product Code. 16865413
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Multi-faceted polygon shape with increased surface area creates added turbulence relative to similar sized, smooth cylindrical bars to provide thorough mixing
  • Absence of a pivot ring allows the Spinbar to stay close to the bottom of the vessel enabling “scraping” of solids
  • Optimal coupling with magnetic spindrive as entire length of stir bar is in close contact
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE Coating; - Superior Chemical Compatibility
  • Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
  • Autoclave safe
Specifications
Specifications
Autoclavable
PTFE, FDA-grade, USP Class VI
38 mm
10 Pk.
White
10 mm
PTFE
Certificates
Promotions
Promotions
Product Content Correction
Product Title
SP Bel-Art™ Metric Polygon Spinbar Magnetic Stirring Bars without Pivot Ring > 38 x 10 mm