Learn More
SP Bel-Art™ Metric Polygon Spinbar Magnetic Stirring Bars without Pivot Ring
High quality PTFE encapsulated magnetic stirring bars ensure efficient mixing with consistent results.
Brand:  SP Bel-Art™ F37120-6007
 View more versions of this product
Product Code. 16895413
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
  • Multi-faceted polygon shape with increased surface area creates added turbulence relative to similar sized, smooth cylindrical bars to provide thorough mixing
  • Absence of a pivot ring allows the Spinbar to stay close to the bottom of the vessel enabling “scraping” of solids
  • Optimal coupling with magnetic spindrive as entire length of stir bar is in close contact
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE Coating; - Superior Chemical Compatibility
  • Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
  • Autoclave safe
Specifications
Specifications
Autoclavable
PTFE, FDA-grade, USP Class VI
60 mm
5 Pk.
White
7 mm
PTFE
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SP Bel-Art™ Metric Polygon Spinbar Magnetic Stirring Bars without Pivot Ring > 60 x 7 mm