Learn More
Midkine Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28546-1-AP-20UL
Description
The Midkine antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Midkine. This antibody recognizes human, mouse antigen. The Midkine antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Midkine | |
Polyclonal | |
Unconjugated | |
20 μL | |
4192 | |
IgG |
Western Blot, ELISA | |
233 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |