Learn More
Merck Millex™-FG Syringe Filter Unit, Hydrophobic PTFE, 0.2 μm
Features a hydrophobic Fluoropore™ (PTFE) membrane. Millex™-FG Syringe Filter, 0.20μm, Hydrophobic PTFE provides broad chemical compatibility and is ideal for filtering samples in organic solvents or to prevent back-streaming of atmospheric moisture in venting applications.
2360.00 SEK - 19580.00 SEK
Specifications
Disposable | Single-use |
---|---|
Material (Membrane) | PTFE (Hydrophobic) |
Pore Size | 0.2 μm |
Housing Material | Polypropylene |
Product Code | Brand | Diameter (Metric) | Connectors | Volume (Metric) | Quantity | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Connectors | Volume (Metric) | Quantity | Price | Quantity & Availability | ||
10094380
|
Merck
SLFG85000 |
50 mm | HB/HB | 100 mL | 100/Pk. |
16850.00 SEK
Pack of 100 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||
10421551
|
Merck
SLFG05010 |
50 mm | HB/HB | 4 L | 10/Pk. |
3930.00 SEK
Pack of 10 |
Estimated Shipment: 08-08-2025
Log in to see stock. |
||
10267770
|
Merck
SLFGN25VS |
25 mm | FLL/MLS with Needle | 100 mL | 25/Pk. |
2360.00 SEK
Pack of 25 |
Estimated Shipment: 29-07-2025
Log in to see stock. |
||
10712911
|
Merck
SLFG05000 |
50 mm | HB/HB | 4 L | 100/Pk. |
15870.00 SEK
Pack of 100 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||
11516515
|
Merck
SLFGL25BS |
25 mm | FLL/MLL | 100 mL | 50/Pk. |
4845.00 SEK
Pack of 50 |
Estimated Shipment: 29-07-2025
Log in to see stock. |
||
10628261
|
Merck
SLFG65000 |
50 mm | HB- 1/8 NPTM | 4 L | 100/Pk. |
19580.00 SEK
Pack of 100 |
Estimated Shipment: 27-08-2025
Log in to see stock. |
||
10410011
|
Merck
SLFG65010 |
50 mm | HB -1/8 NPTM | 4 L | 10/Pk. |
2620.00 SEK
Pack of 10 |
Estimated Shipment: 06-08-2025
Log in to see stock. |
||
10238090
|
Merck
SLFG85010 |
50 mm | HB/HB | 100 mL | 10/Pk. |
2720.00 SEK
Pack of 10 |
N/A | ||
10638261
|
Merck
SLFG75010 |
50 mm | 1/8 NPTM (In/Out) | 100 mL | 10/Pk. |
2525.00 SEK
Pack of 10 |
Estimated Shipment: 29-07-2025
Log in to see stock. |
||
10687121
|
Merck
SLFG55010 |
50 mm | 1/8 NPTM | 4 L | 10/Pk. |
2620.00 SEK
Pack of 10 |
N/A | ||
10648261
|
Merck
SLFG75000 |
50 mm | 1/8 NPTM (In/Out) | 4 L | 100/Pk. |
19580.00 SEK
Pack of 100 |
Estimated Shipment: 27-08-2025
Log in to see stock. |
||
Description
Protect vacuum sources from contamination and moisture damage
Millex™ syringe filter units with hydrophobic Fluoropore™ or Millipore Express™ PLUS PES membrane are ideal for in-line sterilization of gases and venting sterile containers, and filters with Fluoropore™ membrane can also be used for sterilizing or clarifying organic solutions. There are also specialized filter units to protect hemodialysis transducers from blood and moisture. The 50 mm Millex™ filter units are especially useful for vacuum line protection. All units are bi-directional.Features & Benefits:
- Specialized filter units protect hemodialysis transducers from blood and moisture
- 50 mm Millex™ units ideal for vacuum line protection
- All Millex™ Hydrophobic PTFE units are bi-directional in flow
Applications:
Sterilizing Gases, Vacuum Line Protection, Venting Sterile Containers, Sterilizing and Clarifying Organic Solutions
Specifications
Single-use | |
PTFE (Hydrophobic) | |
0.2 μm | |
Polypropylene |