Learn More
Kartell™ MiniFix Micropipettor
An innovative product line of fixed-volume air dispacement mini-micropipettes. Kartell™ MiniFix Micropipettor is designed for fast, repetitive, and precise operation and are competitively priced.
265.00 SEK - 330.72 SEK
Specifications
Certifications/Compliance | CE/IVD |
---|---|
Autoclavable | Autoclavable |
Color | Red |
Number of Channels | 1 |
For Use With (Application) | Routine or biological hazard work, in situ analyses and sampling |
Product Code | Brand | Volume (Metric) | Compatible Tips | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Compatible Tips | Price | Quantity & Availability | |||||
10798293
|
Kartell™
0129000 |
100 μL | Kartell™ 960/958 |
293.00 SEK
Each |
Estimated Shipment: 26-06-2025
Log in to see stock. |
|||||
10257222
|
Kartell™
0128700 |
20 μL | Kartell™ 960/958 |
294.00 SEK
Each |
Estimated Shipment: 26-06-2025
Log in to see stock. |
|||||
10181473
|
Kartell™
0129300 |
500 μL | Kartell™ 961 |
265.00 SEK
Each |
Estimated Shipment: 26-06-2025
Log in to see stock. |
|||||
10790404
|
Kartell™
0128600 |
10 μL | Kartell™ 960/958 |
265.00 SEK
Each |
Estimated Shipment: 26-06-2025
Log in to see stock. |
|||||
10053702
|
Kartell™
0128800 |
25 μL | Kartell™ 960/958 |
330.72 SEK
Each |
Estimated Shipment: 03-07-2025
Log in to see stock. |
|||||
10434242
|
Kartell™
0128500 |
5 μL | Kartell™ 956/959 |
N/A
|
N/A | |||||
10389142
|
Kartell™
0128900 |
50 μL | Kartell™ 960/958 |
265.00 SEK
Each |
Estimated Shipment: 27-06-2025
Log in to see stock. |
|||||
Description
- Fully autoclavable
- Designed for fast, repetitive and precise operation
- Very compact in terms of size and weight
- Designed for routine or biological hazard work
- Kartell™ tips fit on the conical shaft as well as most other commercially available tips
- The plunger button has a different color based on dosing capacity
- Individually calibrated in compliance with ISO8655, the new international standard for piston-operated volumetric apparatus
- Carries the CE conformity mark
Specifications
CE/IVD | |
Red | |
Routine or biological hazard work, in situ analyses and sampling | |
13 cm |
Autoclavable | |
1 | |
1/Pk. | |
10 g |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Kartell™ MiniFix Micropipettor