Learn More
VistaLab™ Technologies MLA™ Silver Brand Precision Pipetters
Made of precision engineered aluminum alloy and stainless steel with limited Lifetime Warranty (covering parts). Individually serialized with Calibration Certificate included.
Brand: VistaLab™ Technologies 1107
This item is not returnable.
View return policy
Description
- 'To deliver' capability ensures volume accuracy over a wide range of viscosities
- Ideal for repetitive delivery of reagents when high precision and accuracy are required
- Capacity engraved on bonnet
Specifications
±2% | |
Silver | |
Micropipette Tip | |
MLA Precision | |
Non-sterile | |
70 ÎĽL | |
±0.57% | |
Limited Lifetime | |
2 in. |
Not Autoclavable | |
Reusable | |
Aluminum Alloy, Stainless Steel | |
1 | |
Precision Pipetter | |
Nozzle Insert, Calibration Key | |
1 Ea. | |
9 in. | |
1.5 in. |
Product Content Correction
Product Title
VistaLab™ Technologies MLA™ Silver Brand Precision Pipetters > Volume: 70 uL
Spot an opportunity for improvement?Share a Content Correction