missing translation for 'onlineSavingsMsg'
Learn More

MNT Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 23742-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16882425

  • 1355.00 SEK / 20µL
Estimated Shipment: 28-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Myc/Max/Mad network comprises a group of transcription factors that co-interact to regulate gene-specific transcriptional activation or repression. This gene encodes a protein member of the Myc/Max/Mad network. This protein has a basic-Helix-Loop-Helix-zipper domain (bHLHzip) with which it binds the canonical DNA sequence CANNTG, known as the E box, following heterodimerization with Max proteins. This protein is likely a transcriptional repressor and an antagonist of Myc-dependent transcriptional activation and cell growth. This protein represses transcription by binding to DNA binding proteins at its N-terminal Sin3-interaction domain.
Specifications

Specifications

MNT
Polyclonal
Unconjugated
MNT
bHLHd3, MAD6, MAX binding protein, Max binding protein MNT, MNT, MXD6, Myc antagonist MNT, Protein ROX, ROX
Rabbit
Antigen Affinity Chromatography
RUO
4335
-20°C
Liquid
Immunofluorescence, Western Blot, Immunocytochemistry
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q99583
MNT
MNT Fusion Protein Ag20666
20 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers