Learn More
Hamilton™ 1700 Series Standard PTFE Instrument Syringe
Setting the standard for quality and reliability.
Brand:  Hamilton™ 5495-10
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 10097109
  • 2350.00 SEK / Each
Estimated Shipment: 31-05-2024
to see stock.
Description
Description
  • Syringe life is exceptional when used in conjunction with Hamilton™ precision syringe drives
  • Enables precision liquid handling
  • Gastight for leak-free performance
Specifications
Specifications
12.5 μL