missing translation for 'onlineSavingsMsg'
Learn More
Propargylamine, 99% Product Code.: 15204634

Propargylamine, 99%

Product Code. 15204634 Shop All Thermo Scientific Chemicals Products
1 kg, 1kg
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Unit Size:
100g
1kg
25g
5g
This item is not returnable. View return policy

Product Code. 15204634

Brand: Thermo Scientific Acros 131460010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2450-71-7 | C3H5N | 55.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2450-71-7
Molecular Formula C3H5N
Molecular Weight (g/mol) 55.08
MDL Number MFCD00008198
InChI Key JKANAVGODYYCQF-UHFFFAOYSA-N
Synonym propargylamine, 2-propynylamine, 2-propyn-1-amine, 3-aminopropyne, 3-amino-1-propyne, 2-propyn-1-thiol, 3-aminoprop-1-yne, propargylamin, pro-pargylamine, propargyl amine
PubChem CID 239041
IUPAC Name prop-2-yn-1-amine
SMILES C#CCN

Specifications

Color Colorless to Yellow
Density 0.8600g/mL
Boiling Point 84.0°C
Flash Point 3°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Linear Formula HCidentical toCCH2NH2
Refractive Index 1.4460 to 1.4500
Quantity 1 kg
Beilstein 04,228
Specific Gravity 0.86
Solubility Information Solubility in water: miscible in water.
Formula Weight 55.08
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Propargylamine, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberA0473832 Certificate TypeCertificate of Analysis Date02/07/2025 Product Code131460010,131461000,131460250,131460050
Lot NumberA0452687 Certificate TypeCertificate of Analysis Date25/06/2025 Product Code131460010,131461000,131460250,131460050
Lot NumberA0470889 Certificate TypeCertificate of Analysis Date17/05/2025 Product Code131460010,131461000,131460250,131460050
Lot NumberA0447087 Certificate TypeCertificate of Analysis Date27/11/2023 Product Code131460010,131461000,131460250,131460050
Lot NumberA0452687 Certificate TypeCertificate of Analysis Date25/08/2023 Product Code131460010,131461000,131460250,131460050

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Propargylamine, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.