Learn More
Mouse NTB-A/SLAMF6 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB3986-SP
This item is not returnable.
View return policy
Description
NTB-A/SLAMF6/CD352 Monoclonal specifically detects NTB-A/SLAMF6/CD352 in Mouse samples. It is validated for Western Blot.
Specifications
NTB-A/SLAMF6/CD352 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q9ET39 | |
SLAMF6 | |
Mouse myeloma cell line NS0-derived recombinant mouse NTB-A/SLAMF6 Glu31-Asn239 Accession # Q9ET39 | |
25 μg | |
Primary | |
Detects mouse NTB-A/SLAMF6 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human NTB-A or recombinant mouse CD84 is observed. | |
Mouse | |
Purified |
Western Blot | |
448318 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Activating NK receptor, CD352, CD352 antigen, KALIFLJ50657, Ly108, NK-T-B-antigen, NTBA, NTBA receptor, NTB-AMGC104953, NTBAT- and B-cell antigen, SF2000, SLAM family member 6, SLAMF6 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
114836 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Product Title
Mouse NTB-A/SLAMF6 Antibody, R&D Systems™ > 25μg; Unlabeled
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction