missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse NTB-A/SLAMF6 Antibody, R&D Systems™
Rat Monoclonal Antibody
1875.00 SEK - 4095.00 SEK
Specifications
Antigen | NTB-A/SLAMF6/CD352 |
---|---|
Clone | 448318 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18702048
|
R&D Systems
MAB3986-SP |
25 ÎĽg |
1875.00 SEK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18182411
|
R&D Systems
MAB3986 |
100 ÎĽg |
4095.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NTB-A/SLAMF6/CD352 Monoclonal specifically detects NTB-A/SLAMF6/CD352 in Mouse samples. It is validated for Western Blot.Specifications
NTB-A/SLAMF6/CD352 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
Q9ET39 | |
114836 | |
Mouse myeloma cell line NS0-derived recombinant mouse NTB-A/SLAMF6 Glu31-Asn239 Accession # Q9ET39 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse NTB-A/SLAMF6 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human NTB-A or recombinant mouse CD84 is observed. |
448318 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 ÎĽm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 ÎĽm filtered solution in PBS. with No Preservative | |
Activating NK receptor, CD352, CD352 antigen, KALIFLJ50657, Ly108, NK-T-B-antigen, NTBA, NTBA receptor, NTB-AMGC104953, NTBAT- and B-cell antigen, SF2000, SLAM family member 6, SLAMF6 | |
SLAMF6 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Mouse NTB-A/SLAMF6 Antibody, R&D Systems™