Learn More
STEMCELL Technologies Mouse Recombinant IL-4
Interleukin 4
Brand:  STEMCELL Technologies 78047.2
Encompass_Preferred
 View more versions of this product
Product Code. 17199431
  • 42785.55 SEK / 1000µg
This item is not returnable. View return policy
Description
Description
  • Interleukin 4 (IL-4) is a type I cytokine produced by Th2 T cells, NK T cells, γ/δ T cells, eosinophils, mast cells, and activated basophils
  • IL-4 receptors are present on a number of cells, including hematopoietic, endothelial, epithelial, muscle, and brain tissues
  • IL-4 receptor engagement initiates signaling through the JAK/STAT pathway, and leads to the activation of PI3K and Ras/MAPK pathways (Nelms et al.) IL-4 plays a key role in immunoglobulin class switching in B cells and in regulating the differentiation of naïve T cells into T helper 2 (Th2) cells, while inhibiting Th1 differentiation
  • IL-4 acts together with tumor necrosis factor to stimulate expression of adhesion molecules on vascular endothelial cells and to downregulate expression of E-selectin, thus recruiting T cells and eosinophils to the site of inflammation (Nelms et al.)
Specifications
Specifications
B cell stimulating factor, BCDF, BCGF, BSF-1, Interleukin-4
1000 μg
B Cells; Dendritic Cells; Hematopoietic Stem and Progenitor Cells; Lymphocytes; Monocytes; Myeloid Cells; Neurons; T Cells
≥95%
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
STEMCELL Technologies Mouse Recombinant IL-4