missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Nigrosin Stain
Used as a viable cell staining agent especially in stem cell biology
583.00 SEK - 1934.50 SEK
Specifications
Type | Nigrosin |
---|---|
Format | Crystal |
For Use With (Application) | Viable cell staining agent especially in stem cell biology |
Storage Requirements | 15 to 30°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11454992
|
MP Biomedicals™
0219459380 |
100 g |
1934.50 SEK
100g |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
11444992
|
MP Biomedicals™
0219459325 |
25 g |
583.00 SEK
25g |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
Description
- Formula Weight: 737.87
- CAS # 8005-03-6
- Synonym: Acid Black 2; C.I. 50420
- Physical Description: Black crystalline powder
- Solublility: Soluble in water(1mg/ml-opaque black with purple cast solution)
Specifications
Nigrosin | |
Crystal | |
Viable cell staining agent especially in stem cell biology | |
15 to 30°C |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.