missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPP8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16796-1-AP-150UL
This item is not returnable.
View return policy
Description
Heterochromatin component that specifically recognizes and binds methylated ′Lys-9′ of histone H3 (H3K9me) and promotes recruitment of proteins that mediate epigenetic repression. Mediates recruitment of the HUSH complex to H3K9me3 sites: the HUSH complex is recruited to genomic loci rich in H3K9me3 and is probably required to maintain transcriptional silencing by promoting recruitment of SETDB1, a histone methyltransferase that mediates further deposition of H3K9me3. Binds H3K9me and promotes DNA methylation by recruiting DNMT3A to target CpG sites; these can be situated within the coding region of the gene. Mediates down-regulation of CDH1 expression.Specifications
MPP8 | |
Polyclonal | |
Unconjugated | |
MPHOSPH8 | |
HSMPP8, M phase phosphoprotein 8, MPHOSPH8, MPP8, TWA3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
290270, 54737, 75339 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
G3V8T1, Q3TYA6, Q99549 | |
Mphosph8 | |
MPP8 Fusion Protein Ag10268 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MPP8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction