missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTERF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16957-1-AP-20UL
This item is not returnable.
View return policy
Description
Transcription termination factor. Binds to a 28 bp region within the tRNA(Leu(uur)) gene at a position immediately adjacent to and downstream of the 16S rRNA gene; this region comprises a tridecamer sequence critical for directing accurate termination. Binds DNA along the major grove and promotes DNA bending and partial unwinding. Promotes base flipping. Transcription termination activity appears to be polarized with highest specificity for transcripts initiated on the light strand.Specifications
MTERF | |
Polyclonal | |
Unconjugated | |
MTERF1 | |
MTERF, MTERF1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
545725, 7978, 85261 | |
-20°C | |
Liquid |
Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8CHZ9, Q99551, Q9EPI8 | |
Mterf1, Mterf1a | |
MTERF Fusion Protein Ag10640 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MTERF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction