missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTERF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | MTERF |
---|---|
Concentration | 0.29 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16896754
|
Proteintech
16957-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16886754
|
Proteintech
16957-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Transcription termination factor. Binds to a 28 bp region within the tRNA(Leu(uur)) gene at a position immediately adjacent to and downstream of the 16S rRNA gene; this region comprises a tridecamer sequence critical for directing accurate termination. Binds DNA along the major grove and promotes DNA bending and partial unwinding. Promotes base flipping. Transcription termination activity appears to be polarized with highest specificity for transcripts initiated on the light strand.Specifications
MTERF | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
Q8CHZ9, Q99551, Q9EPI8 | |
545725, 7978, 85261 | |
MTERF Fusion Protein Ag10640 | |
Primary | |
-20°C | |
MTERF1 |
0.29 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
MTERF, MTERF1 | |
Mterf1, Mterf1a | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MTERF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech