Learn More
Merck MultiScreen 96-Well Collection Plates
MultiScreen™ HTS plates are fully compatible with automation
1925.00 SEK - 10795.00 SEK
Specifications
Format Standard
Array 8 x 12
No. of Wells 96
Well Color Clear
View More Specs
Products 2
Product Code Brand Material Price Quantity & Availability  
Product Code Brand Material Price Quantity & Availability  
10727284
View Documents
Merck
MSCPNPS00
Polystyrene
1925.00 SEK
Pack of 100
Only null left
Estimated Shipment: 06-08-2025
Add to basket
10205993
View Documents
Merck
MSCPNUV40
Polypropylene
10795.00 SEK
Pack of 40
Only null left
Estimated Shipment: 25-08-2025
Add to basket
Description
Description
Fully Compatible with Automation
High throughput MultiScreen™HTS filter plates are specifically developed for use with automation. The 96 well and 384 well plates are designed to standardised dimensions and meet ANSI/SBS 2004 plate specifications. They are fully compatible with automated gripper arms, stackers, barcode labels, and plate readers.
Innovative Plate Design
Rigid sidewalls are aligned for easy handling by robotic gripper arm and also provide ample surfaces for bar code labels. The plate design protects the underdrain and eliminates surface contact with individual well 'drip directors'. The plastic skirt enables stacking directly with collection plates and improves the vacuum seal when used with a Millipore™ vacuum manifold. The removable underdrain gives full access to membranes and is ideal for applications including Elispot, radioisotope detection and whole organism visualisation.
Extensive Product Range
MultiScreen™HTS filter plates are available in a broad range of Millipore plastics and membranes. MultiScreen™ Solubility filter plates are also optimised in HTS format for filtration-based solubility determination. Accessory column loaders are available for loading chromatography media for column separators. MultiScreen™HTS filter plates are available in clear, opaque and white. For non-standard plastic and membrane combinations, custom requests are accepted. Available membranes include:
  • Hydrophilic Durapore™ PVDF membrane
  • Glass fibre
  • Hydrophobic Immobilon™-P membrane
  • HA mixed cellulose esters
  • Negatively charged phosphocellulose
  • Positively charged DEAE
  • Track-etched polycarbonate Applications include: receptor-ligand binding; general sample preparation; Elispot; enzyme activity; bead-based assays; whole organism visualisation and radioisotope detection.
Specifications
Specifications
Standard
8 x 12
96
Clear
Documents
Product Certifications
Product Content Correction
Product Title
Merck MultiScreen 96-Well Collection Plates