missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYO1E Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17768-1-AP-150UL
This item is not returnable.
View return policy
Description
Myosins are actin-based motor molecules with ATPase activity. Unconventional myosins serve in intracellular movements. Their highly divergent tails are presumed to bind to membranous compartments, which would be moved relative to actin filaments.Specifications
MYO1E | |
Polyclonal | |
Unconjugated | |
MYO1E | |
HuncM IC, MYO1C, MYO1E, Myosin Ic, myosin IE | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
25484, 4643, 71602 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
E9Q634, Q12965, Q63356 | |
MYO1E | |
MYO1E Fusion Protein Ag12176 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MYO1E Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction