missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYO1E Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | MYO1E |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16897734
|
Proteintech
17768-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16887734
|
Proteintech
17768-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Myosins are actin-based motor molecules with ATPase activity. Unconventional myosins serve in intracellular movements. Their highly divergent tails are presumed to bind to membranous compartments, which would be moved relative to actin filaments.Specifications
MYO1E | |
Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
E9Q634, Q12965, Q63356 | |
25484, 4643, 71602 | |
MYO1E Fusion Protein Ag12176 | |
Primary | |
-20°C | |
MYO1E |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
HuncM IC, MYO1C, MYO1E, Myosin Ic, myosin IE | |
MYO1E | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MYO1E Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech