missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-dodecane, >99%, MP Biomedicals™
Shop All MP Biomedical Products
Click to view available options
Quantity:
25 mL
100 mL
500 mL
1 L
Unit Size:
1 L
100mL
25mL
500mL
Specifications
Specifications
Color | Colorless |
Density | 0.753 at 25°C |
Assay Percent Range | >99% |
Quantity | 500 mL |
Formula Weight | 170.34 |
Percent Purity | 0.995 |
Physical Form | Liquid |
Chemical Name or Material | N-dodecane |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
N-dodecane, >99%, MP Biomedicals™ >
Spot an opportunity for improvement?Share a Content Correction