Learn More
n-Heptane, for HPLC, (approx. 99% n-Heptane) Product Code.: 10232332
n-Heptane, for HPLC, (approx. 99% n-Heptane)
Product Code. p-2716200 Shop All Thermo Scientific Chemicals Products
1 L, 1 L
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.50 L
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10232332
Brand: Thermo Scientific Acros 364970010
Spotlight
Use promo code 24156 at checkout
View Available Promotions
Request Bulk or Custom Format
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
CAS: 142-82-5 | C7H16 | 100.21 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
Chemical Identifiers
CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC
Specifications
Melting Point -91.0°C
Absorbance (1 cm cell vs HPLC−grade water), (1 cm cell vs HPLC−grade water) at 0nm, 0.005 max. at 250nm, 0.01 max. at 240nm, 0.02 max. at 230nm, 0.05 max. at 220nm, 0.1 max. at 210nm, 0.25 max. at 200nm
Color Colorless
Acidity 0.00015 meq/g max.
Density 0.6830g/mL
Boiling Point 98.0°C
Quantity 1 L
Flash Point −4°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3850 to 1.3890
Residue after Evaporation 5ppm max.
Specific Gravity 0.683
Linear Formula CH3(CH2)5CH3
Viscosity 0.4 mPa.s (20°C)
Beilstein 01, 154
Water 0.01% Max.
Merck Index 15, 4694
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, ether and chloroform
Formula Weight 100.2
Percent Purity ≥99% (GC)
Grade HPLC
Chemical Name or Material n-Heptane
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2540328 Certificate TypeCertificate of Analysis Date11/06/2025 Catalog Number364970025,364970010
Lot Number2527159 Certificate TypeCertificate of Analysis Date17/05/2025 Catalog Number364970025,364970010
Lot Number2527346 Certificate TypeCertificate of Analysis Date17/05/2025 Catalog Number364970025,364970010
Lot Number2527693 Certificate TypeCertificate of Analysis Date17/05/2025 Catalog Number364970025,364970010
Lot Number2530376 Certificate TypeCertificate of Analysis Date17/05/2025 Catalog Number364970025,364970010
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Heptane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P273-Avoid release to the environment.
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24156
    Expires2025-06-30
    Days
    14
    Hours
    7
    Minutes
    50
    Seconds
    13
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/05/2025 and 30/06/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
  • Multiple Manufacturers
    Save up to 59% on some of our most popular products
    Promo Code: 24140
    Expires2025-08-24
    Days
    69
    Hours
    7
    Minutes
    50
    Seconds
    13
RUO – Research Use Only
Product Content Correction
Product Title
n-Heptane, for HPLC, (approx. 99% n-Heptane) >