Learn More
NCOA3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20032-1-AP-20UL
 View more versions of this product
Product Code. 16879514
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
The antibody can recognizes all the isoforms of NCOA3.
AIB1-AC3 recognizes AIB1, a member of the SRC-1 family of nuclear receptor coactivators with an apparent molecular weight of 160kDa. The AIB1 gene has been mapped to region 20q12 and has been shown to be amplified in breast tumors and pancreatic cancer cell lines. AIB1 interacts in a ligand-dependent fashion with the estrogen receptor to enhance estrogen-dependent transcriptional gene activation.
Specifications
Show More
Certificates
Promotions
Promotions
Product Content Correction
Product Title
NCOA3 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated