missing translation for 'onlineSavingsMsg'
Learn More

NCOA3 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1395.65 SEK - 3774.95 SEK

Specifications

Antigen NCOA3
Concentration 0.27 mg/mL
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16879514
View Documents
Proteintech
20032-1-AP-20UL
20 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16869514
View Documents
Proteintech
20032-1-AP-150UL
150 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

The antibody can recognizes all the isoforms of NCOA3.

AIB1-AC3 recognizes AIB1, a member of the SRC-1 family of nuclear receptor coactivators with an apparent molecular weight of 160kDa. The AIB1 gene has been mapped to region 20q12 and has been shown to be amplified in breast tumors and pancreatic cancer cell lines. AIB1 interacts in a ligand-dependent fashion with the estrogen receptor to enhance estrogen-dependent transcriptional gene activation.
Specifications

Specifications

NCOA3
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry, Western Blot
Unconjugated
Rabbit
Human
Q9Y6Q9
8202
Peptide
Primary
-20°C
NCOA3
0.27 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
ACTR, AIB 1, AIB1, CBP interacting protein, KAT13B, NCoA 3, NCOA3, nuclear receptor coactivator 3, pCIP, SRC 3, SRC3, TRAM 1, TRAM1
NCOA3
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
NCOA3 Rabbit anti-Human, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.