Learn More
NDC80 Mouse anti-Human, Unconjugated, Clone: 2B7A8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66960-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The NDC80 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NDC80. This antibody recognizes human, rat, mouse antigen. The NDC80 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
NDC80 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Mouse, Rat, Human |
ELISA, Western Blot | |
2B7A8 | |
Unconjugated | |
20 μL | |
10403 | |
IgG1 |