Learn More
NDC80 Mouse anti-Human, Unconjugated, Clone: 2B7A8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66960-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16879285
  • 3665.00 SEK / 150µL
Estimated Shipment: 03-04-2023
to see stock.
Description
Description
The NDC80 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NDC80. This antibody recognizes human, rat, mouse antigen. The NDC80 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
NDC80
Monoclonal
1000 μg/mL
Mouse
Primary
Mouse, Rat, Human
ELISA, Western Blot
2B7A8
Unconjugated
150 μL
10403
IgG1