missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NeutraKine FGF-4 Mouse anti-Human, Unconjugated, Clone: 1A2D8, Proteintech
Mouse Monoclonal Antibody
1690.93 SEK - 3620.19 SEK
Specifications
Antigen | NeutraKine FGF-4 |
---|---|
Clone | 1A2D8 |
Concentration | 1000 ÎĽg/mL |
Applications | Neutralization, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17224656
|
Proteintech
69022-1-IG-25UG |
25 ÎĽg |
1690.93 SEK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17214656
|
Proteintech
69022-1-IG-100UG |
100 ÎĽg |
3620.19 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody is used to Block/Neutralize FGF-4 protein.
Specifications
NeutraKine FGF-4 | |
1000 ÎĽg/mL | |
Monoclonal | |
Liquid | |
2249 | |
Primary |
1A2D8 | |
Neutralization, ELISA | |
Unconjugated | |
Mouse | |
IgG1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NeutraKine FGF-4 Mouse anti-Human, Unconjugated, Clone: 1A2D8, Proteintech