Learn More
NeutraKine FGF-4 Mouse anti-Human, Unconjugated, Clone: 1A2D8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 69022-1-IG-25UG
This item is not returnable.
View return policy
Description
This antibody is used to Block/Neutralize FGF-4 protein.
Specifications
NeutraKine FGF-4 | |
Monoclonal | |
1000 ÎĽg/mL | |
Mouse | |
Primary | |
Human, Mouse | |
IgG1 |
Neutralization, ELISA | |
1A2D8 | |
Unconjugated | |
25 ÎĽg | |
2249 | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NeutraKine FGF-4 Mouse anti-Human, Unconjugated, Clone: 1A2D8, Proteintech > 25 ÎĽg
Spot an opportunity for improvement?Share a Content Correction