Learn More
NeutraKine™ IFN Alpha 2A Mouse anti-Human, Unconjugated, Clone: 2H8G1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 69008-1-IG-100UG
Description
The NeutraKine™ IFN Alpha 2A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NeutraKine™ IFN Alpha 2A. This antibody recognizes human antigen. The NeutraKine™ IFN Alpha 2A antibody has been validated for the following applications: Neutralization, ELISA analysis.
Specifications
NeutraKine™ IFN Alpha 2A | |
Monoclonal | |
Unconjugated | |
100 ÎĽg | |
3440 | |
IgG1 |
Neutralization | |
2H8G1 | |
Mouse | |
Primary | |
Human |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.