Learn More
NeutraKine IL-17A Mouse anti-Human, Unconjugated, Clone: 1F3E3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 69021-1-IG-100UG
Description
IL17A, also named as IL-17, is a proinflammatory cytokine. IL-17, synthesized only by memory T cells and natural killer cells, has pleiotropic effects, mainly in the recruitment and activation of neutrophils. This cytokine regulates the activities of NF-kappaB and mitogen-activated protein kinases. This cytokine can stimulate the expression of IL6 and cyclooxygenase-2 (PTGS2/COX-2), as well as enhance the production of nitric oxide (NO). High levels of this cytokine are associated with several chronic inflammatory diseases including rheumatoid arthritis, psoriasis and multiple sclerosis. The IL-17 receptor is a type I transmembrane protein, that is widely expressed on epithelial cells, fibroblasts, B and T cells, and monocytic cells. In psoriatic skin lesions, both Th17 cells and their downstream effector molecules, e.g. IL-17 and IL-22, are highly increased. This antibody is used to Block/Neutralize IL-17A protein.
Specifications
NeutraKine IL-17A | |
Monoclonal | |
Unconjugated | |
100 ÎĽg | |
3605 | |
Liquid |
Neutralization, ELISA | |
1F3E3 | |
Mouse | |
Primary | |
Human | |
IgG1 |