Learn More
NeutraKine™ IL-4 Mouse anti-Human, Unconjugated, Clone: 1H5G5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 69005-1-IG-25UG
 View more versions of this product
Product Code. 16840416
  • 1690.93 SEK / 25µg
This item is not returnable. View return policy
Description
Description
The NeutraKine™ IL-4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NeutraKine™ IL-4. This antibody recognizes human antigen. The NeutraKine™ IL-4 antibody has been validated for the following applications: Neutralization, ELISA analysis.
Specifications
Specifications
NeutraKine™ IL-4
Monoclonal
Unconjugated
25 ÎĽg
3565
IgG1
Neutralization
1H5G5
Mouse
Primary
Human
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NeutraKine™ IL-4 Mouse anti-Human, Unconjugated, Clone: 1H5G5, Proteintech > 25 μg