Learn More
NeutraKine™ IL-4 Mouse anti-Human, Unconjugated, Clone: 1H5G5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 69005-1-IG-25UG
 View more versions of this product
Product Code. 16840416
  • 1690.93 SEK / 25µg
Description
The NeutraKine™ IL-4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NeutraKine™ IL-4. This antibody recognizes human antigen. The NeutraKine™ IL-4 antibody has been validated for the following applications: Neutralization, ELISA analysis.
Specifications
Specifications
NeutraKine™ IL-4
Monoclonal
Unconjugated
25 ÎĽg
3565
IgG1
Neutralization
1H5G5
Mouse
Primary
Human
Certificates
Promotions
Promotions
Product Content Correction
Product Title
NeutraKine™ IL-4 Mouse anti-Human, Unconjugated, Clone: 1H5G5, Proteintech > 25 μg