missing translation for 'onlineSavingsMsg'
Learn More

Elma™ Noise Protection Box

Reduce the typical cavitation noise of an operating ultrasonic unit to a tolerable level. Elma™ Noise Protection Box reduces the amount of noise by 20dBA or approximately 75%. For Ultrasonic Water Baths.

Brand:  Elma™ 1033905

Additional Details : Weight : 11.65500kg

 View more versions of this product

Product Code. 10352055

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Perceived noise reduction of 75% allows operation of ultrasonic cleaning units in sensitive work areas, such as chemical and dental laboratories, medical practices or even workshops
  • Vapors are carried off by an exhaust device to protect the box inside against an accumulation of heat or humidity
  • Flap made of plexiglass to allow monitoring of operating process and filling level without opening the noise protection box
  • Noise Protection Box M is for units up to S 30 for operation with glass beakers, test tube rack, Erlenmeyer flasks
  • Noise protection box L is for units up to S 100 for operation with glass beakers, test tube rack, Erlenmeyer flasks
  • 230V
Specifications

Specifications

1 Each
Ultrasonic Cleaner
Plexiglass flap with handle, built-in exhaust
415 x 465 mm
465 mm
Medium
415 mm
360 mm
415 mm
12.6 kg
465 x 415 x 360 mm (WxDxH)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers