missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20751-1-AP-20UL
This item is not returnable.
View return policy
Description
Western blot analysis using this antibody detected a major band around 45-55 kDa in kidney tissue.
The function of this protein remains unknown.Specifications
NPT1 | |
Polyclonal | |
Unconjugated | |
SLC17A1 | |
Na(+)/PI cotransporter 1, Na/Pi 4, NAPI 1, SLC17A1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
171080, 20504, 6568 | |
-20°C | |
Liquid |
Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q14916, Q61983, Q62795 | |
Slc17a1 | |
NPT1 Fusion Protein Ag12819 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NPT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction